N-(Isobutoxymethyl)acrylamide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

I1124-100ML I1124-25ML
TCIAI1124-100MLEA 419 DKK
TCIAI1124-100ML TCIAI1124-25ML
N-(Isobutoxymethyl)acrylamide ≥98.0% (by titrimetric analysis)
N-(Isobutoxymethyl)acrylamide
Formel: C₈H₁₅NO₂
Kogepunkt: 100°C
Flammepunkt: 97°C
MDL Number: MFCD00043107
CAS-nummer: 16669-59-3
EINECS: 240-715-7
UN nummer: 2810
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Bromination Method) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR