Ingenol ≥99.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
I1112-10MG
I1112-50MG
TCIAI1112-10MGEA
632
DKK
TCIAI1112-10MG
TCIAI1112-50MG
Ingenol ≥99.0% (by HPLC)
Ingenol
Formel:
C₂₀H₂₈O₅ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD27980538 CAS-nummer: 30220-46-3 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Melting point | 151.0 to 155.0 deg-C |
Specific rotation [a]20/D | -25.0 to -30.0 deg (C=1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...