Isopropamide iodide ≥95.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: (3-Carbamoyl-3,3-diphenylpropyl)diisopropylmethylammonium iodide, 4-Amino-N,N-diisopropyl-N-methyl-4-oxo-3,3-diphenyl-1-butanaminium iodide
I1048-1G I1048-5G
TCIAI1048-1GEA 1560 DKK
TCIAI1048-1G TCIAI1048-5G
Isopropamide iodide ≥95.0% (by HPLC, titration analysis)
Isopropamide iodide
Formel: C₂₃H₃₃IN₂O
Molekylvægt: 480.43 g/mol
Smeltepunkt: 201°C
MDL Number: MFCD00063513
CAS-nummer: 71-81-8
EINECS: 200-766-8
Merck Index: 14,05202
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0be0a7be50b32fea16e052ac8f74821296dde45c&vid=0be0a7be50b32fea16e052ac8f74821296dde45c&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 95.0 %