Isopropylamine hydrobromide ≥97.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Isopropylammonium bromide

I1041-1G I1041-5G
TCIAI1041-1GEA 132 DKK
TCIAI1041-1G TCIAI1041-5G
Isopropylamine hydrobromide ≥97.0% (by total nitrogen and titration analysis)
Isopropylamine hydrobromide
Formel: C₃H₉N·HBr
Molekylvægt: 140.02 g/mol
MDL Number: MFCD22419143
CAS-nummer: 29552-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4707d335a9dcea49bad1a6f1d83312399d376b27&vid=4707d335a9dcea49bad1a6f1d83312399d376b27&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR