1-Amino-2-(isopropylsulphonyl)benzene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

I1033-1G I1033-5G
TCIAI1033-1GEA 222 DKK
TCIAI1033-1G TCIAI1033-5G
1-Amino-2-(isopropylsulphonyl)benzene ≥98.0% (by GC)
Formel: C₉H₁₃NO₂S
MDL Number: MFCD08445603
CAS-nummer: 76697-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=644727ebe95f5281164cd08c255fc66b11cb2c10&vid=644727ebe95f5281164cd08c255fc66b11cb2c10&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR