4-Isothiocyanato-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Isothiocyanic acid 4-Cyano-3-(trifluoromethyl)phenyl ester, 4-Cyano-3-(trifluoromethyl)phenyl Isothiocyanate

I0987-1G I0987-5G
TCIAI0987-1GEA 476 DKK
TCIAI0987-1G TCIAI0987-5G
4-Isothiocyanato-2-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
4-Isothiocyanato-2-(trifluoromethyl)benzonitrile
Formel: C₉H₃F₃N₂S
Molekylvægt: 228.19 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09800709
CAS-nummer: 143782-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f51471fdf149e7612ef676294fc5ddcf014147e&vid=9f51471fdf149e7612ef676294fc5ddcf014147e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR