4-Iododibenzofuran ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Iododibenzo[b,d]furan

I0972-200MG I0972-1G
TCIAI0972-200MGEA 895 DKK
TCIAI0972-200MG TCIAI0972-1G
4-Iododibenzofuran ≥98.0% (by GC)
4-Iododibenzofuran
Formel: C₁₂H₇IO
Molekylvægt: 294.09 g/mol
Smeltepunkt: 73°C
MDL Number: MFCD00094364
CAS-nummer: 65344-26-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07de8ba3f7dbc5f4e26946068a618bbebea93a45&vid=07de8ba3f7dbc5f4e26946068a618bbebea93a45&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR