Imidazole Hydroiodide, low water content ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Imidazolium iodide

I0970-1G I0970-5G
TCIAI0970-1GEA 517 DKK
TCIAI0970-1G TCIAI0970-5G
Imidazole Hydroiodide, low water content ≥98.0% (by titrimetric analysis)
Imidazole Hydroiodide
Formel: C₃H₄N₂·HI
CAS-nummer: 68007-08-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=645a3bbc67a6f2a8b9a2f949241863c43e2c502f&vid=645a3bbc67a6f2a8b9a2f949241863c43e2c502f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR