Isoxazolidine hydrochloride ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

I0966-1G I0966-5G
TCIAI0966-1GEA 838 DKK
TCIAI0966-1G TCIAI0966-5G
Isoxazolidine hydrochloride ≥97.0% (by titrimetric analysis)
Isoxazolidine hydrochloride
Formel: C₃H₇NO·HCl
Molekylvægt: 109.55 g/mol
MDL Number: MFCD11100465
CAS-nummer: 39657-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55be7fd6261467825e8ff23b05397da382ce5159&vid=55be7fd6261467825e8ff23b05397da382ce5159&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR