Iron(III) meso-tetraphenylporphine chloride ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Iron(III) Tetraphenylporphyrin chloride, 5,10,15,20-Tetraphenyl-21H,23H-porphine Iron(III) chloride, meso-Tetraphenylporphyrin iron(III) chloride, iron(III) chloride TPP, 21H,23H-Porphine, 5,10,15,20-tetraphenyl-, iron complex, 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride

I0937-1G I0937-200MG
TCIAI0937-1GEA 1420 DKK
TCIAI0937-1G TCIAI0937-200MG
Iron(III) meso-tetraphenylporphine chloride ≥97.0% (by HPLC, titration analysis)
Iron(III) meso-tetraphenylporphine chloride
Formel: C₄₄H₂₈ClFeN₄
Smeltepunkt: 300°C
MDL Number: MFCD00012152
CAS-nummer: 16456-81-8

Bestil nu

Specifikationer Testresultater

Appearance Green to Dark green to Dark blue powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Chelometric Titration) min. 97.0 %
Molar extinction coefficient min. 100000 (THF, 414 to 418nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR