Imatinib mesylate ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide methanesulphonate, Imatinib Methanesulphonate

I0936-100MG I0936-1G
TCIAI0936-100MGEA 485 DKK
TCIAI0936-100MG TCIAI0936-1G
Imatinib mesylate ≥98.0% (by HPLC)
Imatinib mesylate
Formel: C₂₉H₃₁N₇O·CH₄SO₃
Molekylvægt: 589.72 g/mol
MDL Number: MFCD04307699
CAS-nummer: 220127-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b17449edc2e6bb9fc7c8f1b5c19e823881d78d8c&vid=b17449edc2e6bb9fc7c8f1b5c19e823881d78d8c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR