2-Isocyanatoethyl acrylate ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
I0918-5G
I0918-25G
TCIAI0918-5GEA
542
DKK
TCIAI0918-5G
TCIAI0918-25G
2-Isocyanatoethyl acrylate ≥98.0% (by GC) stabilised
2-Isocyanatoethyl acrylate
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...