4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
BPADA, 4,4'-(4,4'-Isopropylidenediphenoxy)bis(phthalic anhydride), 5,5'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(isobenzofuran-1,3-dione)
I0856-25G
I0856-100G
TCIAI0856-25GEA
394
DKK
TCIAI0856-25G
TCIAI0856-100G
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride ≥98.0% (by HPLC, titration analysis)
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride
Formel:
C₃₁H₂₀O₈ Molekylvægt: 520.49 g/mol |
MDL Number:
MFCD00319167 CAS-nummer: 38103-06-9 EINECS: 253-781-7 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 188.0 to 192.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...