Idebenone ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-1,4-benzoquinone, 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione

I0848-1G I0848-5G I0848-25G
TCIAI0848-1GEA 452 DKK
TCIAI0848-1G TCIAI0848-5G TCIAI0848-25G
Idebenone ≥98.0% (by HPLC, titration analysis)
Idebenone
Formel: C₁₉H₃₀O₅
Molekylvægt: 338.44 g/mol
MDL Number: MFCD00274552
CAS-nummer: 58186-27-9
Merck Index: 14,04888
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fca859b9f7ae8b09a57ae539c3fa4419c033036&vid=8fca859b9f7ae8b09a57ae539c3fa4419c033036&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Iodometric Titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR