Isophorone oxide ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4,4,6-Trimethyl-7-oxabicyclo[4.1.0]heptan-2-one, 2,3-Epoxy-3,5,5-trimethyl-1-cyclohexanone, 3,5,5-Trimethyl-2,3-epoxy-1-cyclohexanone
I0805-25G
I0805-5G
TCIAI0805-25GEA
846
DKK
TCIAI0805-25G
TCIAI0805-5G
Isophorone oxide ≥97.0% (by GC)
Isophorone oxide
Formel:
C₉H₁₄O₂ Molekylvægt: 154.21 g/mol Kogepunkt: 80°C Flammepunkt: 83°C |
MDL Number:
MFCD00085465 CAS-nummer: 10276-21-8 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...