4-Isopropylpiperidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

I0733-1G I0733-5G
TCIAI0733-1GEA 920 DKK
TCIAI0733-1G TCIAI0733-5G
4-Isopropylpiperidine ≥98.0% (by GC, titration analysis)
4-Isopropylpiperidine
Formel: C₈H₁₇N
Kogepunkt: 171°C
MDL Number: MFCD09055342
CAS-nummer: 19678-58-1
UN nummer: 1993
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR