Indole-3-acetamide ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (1H-Indol-3-yl)acetamide

I0668-25G I0668-5G
TCIAI0668-25GEA 1290 DKK
TCIAI0668-25G TCIAI0668-5G
Indole-3-acetamide ≥98.0% (by HPLC, total nitrogen)
Indole-3-acetamide
Formel: C₁₀H₁₀N₂O
MDL Number: MFCD00005641
CAS-nummer: 879-37-8
EINECS: 212-904-4

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 152.0 to 156.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR