3-Isocyanatopropyltriethoxysilane ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
I0556-100G
I0556-25G
TCIAI0556-100GEA
936
DKK
TCIAI0556-100G
TCIAI0556-25G
3-Isocyanatopropyltriethoxysilane ≥95.0% (by GC)
3-Isocyanatopropyltriethoxysilane
Formel:
C₁₀H₂₁NO₄Si Flammepunkt: 77°C |
MDL Number:
MFCD00051459 CAS-nummer: 24801-88-5 EINECS: 246-467-6 UN nummer: 2927 ADR: 6.1,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...