Iodomesitylene diacetate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,4,6-Trimethyl(diacetoxyiodo)benzene, 2,4,6-Trimethyliodobenzene Diacetate, 2-(Diacetoxyiodo)mesitylene

I0479-25G I0479-5G
TCIAI0479-25GEA 1630 DKK
TCIAI0479-25G TCIAI0479-5G
Iodomesitylene diacetate ≥98.0% (by titrimetric analysis)
Iodomesitylene diacetate
Formel: C₁₃H₁₇IO₄
Molekylvægt: 364.18 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00674196
CAS-nummer: 33035-41-5

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR