Isobutylene sulphide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2,2-Dimethylthiirane, 2-Methylpropylene sulphide, 2,2-Dimethylethylene Sulphide

I0461-25ML I0461-5ML
TCIAI0461-25MLEA 2370 DKK
TCIAI0461-25ML TCIAI0461-5ML
Isobutylene sulphide ≥98.0% (by GC)
Isobutylene sulphide
Formel: C₄H₈S
Molekylvægt: 88.17 g/mol
Kogepunkt: 86°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00191662
CAS-nummer: 3772-13-2
UN nummer: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62f147c1955eff78f2f878c02c194a303d1a9862&vid=62f147c1955eff78f2f878c02c194a303d1a9862&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR