3-Iodo-2,4,5-trimethylthiophene ≥96.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Stabilization: Stabilised with copper chip

I0445-1G I0445-5G
TCIAI0445-1GEA 1260 DKK
TCIAI0445-1G TCIAI0445-5G
3-Iodo-2,4,5-trimethylthiophene ≥96.0% (by GC) stabilised
3-Iodo-2,4,5-trimethylthiophene
Formel: C₇H₉IS
Kogepunkt: 87°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00191528
CAS-nummer: 60813-84-5

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR