Isomannide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
I0406-25G
I0406-5G
TCIAI0406-25GEA
813
DKK
TCIAI0406-25G
TCIAI0406-5G
Isomannide ≥98.0% (by GC)
Isomannide
Formel:
C₆H₁₀O₄ Kogepunkt: 150°C |
MDL Number:
MFCD00064818 CAS-nummer: 641-74-7 EINECS: 211-374-1 |
|
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Specific rotation [a]20/D | +85.0 to +95.0 deg (C=3, H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...