p-Tolyl isobutyrate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Isobutyric Acid p-Tolyl Ester

I0396-25ML I0396-5ML
TCIAI0396-25MLEA 2370 DKK
TCIAI0396-25ML TCIAI0396-5ML
p-Tolyl isobutyrate ≥97.0% (by GC)
p-Tolyl isobutyrate
Formel: C₁₁H₁₄O₂
Molekylvægt: 178.23 g/mol
Kogepunkt: 237°C
Flammepunkt: 100°C
MDL Number: MFCD00026440
CAS-nummer: 103-93-5
EINECS: 203-159-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=335bccc61115eea48aa4281d2e54e953e8163362&vid=335bccc61115eea48aa4281d2e54e953e8163362&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR