Isovaleronitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Isobutyl Cyanide

I0292-25ML I0292-500ML I0292-100ML
TCIAI0292-25MLEA 591 DKK
TCIAI0292-25ML TCIAI0292-500ML TCIAI0292-100ML
Isovaleronitrile ≥98.0% (by GC)
Isovaleronitrile
Formel: C₅H₉N
Molekylvægt: 83.13 g/mol
Kogepunkt: 130°C
Flammepunkt: 28°C
MDL Number: MFCD00001944
CAS-nummer: 625-28-5
EINECS: 210-884-1
UN nummer: 1992
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40f07c0743c13aa160dc2a428405f10bb5904ae2&vid=40f07c0743c13aa160dc2a428405f10bb5904ae2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR