Isoproterenol sulphate dihydrate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Isoprenaline Sulfate

I0261-25G I0261-5G
TCIAI0261-25GEA 1970 DKK
TCIAI0261-25G TCIAI0261-5G
Isoproterenol sulphate dihydrate ≥98.0% (by HPLC, total nitrogen)
Isoproterenol sulphate dihydrate
Formel: C₂₂H₃₄N₂O₆·H₂SO₄·2H₂O
Molekylvægt: 520.59 g/mol
MDL Number: MFCD00013139
CAS-nummer: 299-95-6
EINECS: 206-085-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24e8aa76eab4114dc122058ad5699deb3e883feb&vid=ae14a4e1acf8dc76f83f30737df287e0bb16b644&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 6.5 to 7.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR