Isooctadecenylsuccinic anhydride (mixture of branched chain isomers) ≥90.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

I0237-25G I0237-500G
TCIAI0237-25GEA 222 DKK
TCIAI0237-25G TCIAI0237-500G
Isooctadecenylsuccinic anhydride (mixture of branched chain isomers) ≥90.0% (by titrimetric analysis)
Isooctadecenylsuccinic anhydride (mixture of branched chain isomers)
Formel: C₂₂H₃₈O₃
Flammepunkt: 201°C
MDL Number: MFCD00070628
CAS-nummer: 58239-72-8
EINECS: 261-179-0

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (Morpholine Method) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR