Isopropylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Propan-2-amine hydrochloride, 2-Aminopropane Hydrochloride
I0166-25G
I0166-500G
I0166-100G
TCIAI0166-25GEA
132
DKK
TCIAI0166-25G
TCIAI0166-500G
TCIAI0166-100G
Isopropylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Isopropylamine hydrochloride
Formel:
C₃H₉N Molekylvægt: 95.57 g/mol |
MDL Number:
MFCD00050705 CAS-nummer: 15572-56-2 EINECS: 239-629-2 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % (calcd.on anh.substance) |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in Water | almost transparency |
Water | max. 3.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...