Isobutyryl bromide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

I0114-25G I0114-5G
TCIAI0114-25GEA 1680 DKK
TCIAI0114-25G TCIAI0114-5G
Isobutyryl bromide ≥98.0% (by GC)
Isobutyryl bromide
Formel: C₄H₇BrO
Kogepunkt: 117°C
MDL Number: MFCD00059468
CAS-nummer: 2736-37-0
UN nummer: 2920
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR