(-)-Huperzine A ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(−)-Selagine, (5R,9R,E)-5-Amino-11-ethylidene-7-methyl-5,6,9,10-tetrahydro-5,9-methanocycloocta[b]pyridin-2(1H)-one
H1700-10MG
H1700-100MG
TCIAH1700-10MGEA
255
DKK
TCIAH1700-10MG
TCIAH1700-100MG
(-)-Huperzine A ≥98.0% (by HPLC)
(-)-Huperzine A
Formel:
C₁₅H₁₈N₂O Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00171439 CAS-nummer: 102518-79-6 UN nummer: 1544 ADR: 6.1,II Merck Index: 14,04755 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 230.0 to 234.0 deg-C |
Specific rotation [a]20/D | -144 to -155 deg (C=0.5, MeOH) |
Water | max. 3.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...