Hexadecyltriethoxysilan ≥85.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H1685-5ML H1685-25ML
TCIAH1685-5MLEA 394 DKK
TCIAH1685-5ML TCIAH1685-25ML
Hexadecyltriethoxysilan ≥85.0% (by GC)
Hexadecyltriethoxysilane
Formel: C₂₂H₄₈O₃Si
Kogepunkt: 161°C
MDL Number: MFCD00054793
CAS-nummer: 16415-13-7
EINECS: 240-465-9

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR