2-Hexyldecan-1-amine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H1634-1ML H1634-5ML
TCIAH1634-1MLEA 846 DKK
TCIAH1634-1ML TCIAH1634-5ML
2-Hexyldecan-1-amine ≥98.0% (by GC, titration analysis)
2-Hexyldecan-1-amine
Formel: C₁₆H₃₅N
Kogepunkt: 150°C
MDL Number: MFCD30475384
CAS-nummer: 62281-05-4
UN nummer: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8089de72a4b7f377c69d221467143d09b7886402&vid=8089de72a4b7f377c69d221467143d09b7886402&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR