(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulphonium Hexafluoroantimonate ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H1620-25G H1620-5G
TCIAH1620-25GEA 2890 DKK
TCIAH1620-25G TCIAH1620-5G
(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulphonium Hexafluoroantimonate ≥98.0% (by HPLC)
(4-Hydroxyphenyl)methyl(2-methylbenzyl)sulphonium Hexafluoroantimonate
Formel: C₁₅H₁₇F₆OSSb
CAS-nummer: 141651-31-2
UN nummer: 3077
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR