(3-Hydroxyphenyl)acetonitrile ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Hydroxybenzyl Cyanide

H1615-1G H1615-5G
TCIAH1615-1GEA 1190 DKK
TCIAH1615-1G TCIAH1615-5G
(3-Hydroxyphenyl)acetonitrile ≥95.0% (by GC)
(3-Hydroxyphenyl)acetonitrile
Formel: C₈H₇NO
Molekylvægt: 133.15 g/mol
Kogepunkt: 106°C
MDL Number: MFCD03265300
CAS-nummer: 25263-44-9

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR