2-Hydroxy-3-phenoxypropyl acrylate ≥85.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Acrylic acid 2-Hydroxy-3-phenoxypropyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

H1614-25G H1614-500G
TCIAH1614-25GEA 165 DKK
TCIAH1614-25G TCIAH1614-500G
2-Hydroxy-3-phenoxypropyl acrylate ≥85.0% (by GC) stabilised
2-Hydroxy-3-phenoxypropyl acrylate
Formel: C₁₂H₁₄O₄
Molekylvægt: 222.24 g/mol
Flammepunkt: 89°C
MDL Number: MFCD00066300
CAS-nummer: 16969-10-1
EINECS: 241-045-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0907938f90af278dcdfa936bfe449f22a8161344&vid=0907938f90af278dcdfa936bfe449f22a8161344&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR