Hydroxylammoniumchlorid ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Hydroxylamine HCl, Hydroxylamine hydrochloride
H1581-25G
H1581-500G
TCIAH1581-25GEA
148
DKK
TCIAH1581-25G
TCIAH1581-500G
Hydroxylammoniumchlorid ≥97.0% (by titrimetric analysis)
Hydroxylammoniumchlorid
Formel:
NH₂OH·HCl Smeltepunkt: 151°C |
MDL Number:
MFCD00051089 CAS-nummer: 5470-11-1 EINECS: 226-798-2 UN nummer: 2923 ADR: 8,III Merck Index: 14,04828 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Redox method) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...