Flavokawain A ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Flavokawain A, 2'-Hydroxy-4,4',6'-trimethoxychalcone, 1-(2-Hydroxy-4,6-dimethoxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
H1571-1G
H1571-5G
TCIAH1571-1GEA
386
DKK
TCIAH1571-1G
TCIAH1571-5G
Flavokawain A ≥98.0% (by GC)
Flavokawain A
Formel:
C₁₈H₁₈O₅ |
MDL Number:
MFCD00017174 CAS-nummer: 3420-72-2 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 112.0 to 116.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...