2,2,3,4,4,4-Hexafluorobutyl methacrylate ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,2,3,4,4,4-Hexafluorobutyl Methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

H1555-5G H1555-25G H1555-100G
TCIAH1555-5GEA 238 DKK
TCIAH1555-5G TCIAH1555-25G TCIAH1555-100G
2,2,3,4,4,4-Hexafluorobutyl methacrylate ≥98.0% (by GC) stabilised
2,2,3,4,4,4-Hexafluorobutyl methacrylate
Formel: C₈H₈F₆O₂
Flammepunkt: 57°C
MDL Number: MFCD00042311
CAS-nummer: 36405-47-7
UN nummer: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3b244f4c5577088b2c478f0abcd69df96f24ae2&vid=d3b244f4c5577088b2c478f0abcd69df96f24ae2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR