(S)-3-Hydroxypiperidine hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (S)-3-Piperidinol hydrochloride

H1538-1G H1538-5G
TCIAH1538-1GEA 312 DKK
TCIAH1538-1G TCIAH1538-5G
(S)-3-Hydroxypiperidine hydrochloride ≥98.0% (by titrimetric analysis)
(S)-3-Hydroxypiperidine hydrochloride
Formel: C₅H₁₂ClNO
Molekylvægt: 137.6100 g/mol
MDL Number: MFCD03426348
CAS-nummer: 475058-41-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Optical purity (GC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 195.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR