2-(Trifluoromethyl)pyridin-4-ol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Hydroxy-2-(trifluoromethyl)pyridine

H1528-1G H1528-5G
TCIAH1528-1GEA 673 DKK
TCIAH1528-1G TCIAH1528-5G
2-(Trifluoromethyl)pyridin-4-ol ≥98.0% (by GC, titration analysis)
2-(Trifluoromethyl)pyridin-4-ol
Formel: C₆H₄F₃NO
MDL Number: MFCD01571202
CAS-nummer: 170886-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bb8793e9ef973d9d305096637b3a542926908c1&vid=4bb8793e9ef973d9d305096637b3a542926908c1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR