1,1,3,3,5,5-Hexaethoxy-1,3,5-trisilacyclohexane ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H1513-1G H1513-5G
TCIAH1513-1GEA 657 DKK
TCIAH1513-1G TCIAH1513-5G
1,1,3,3,5,5-Hexaethoxy-1,3,5-trisilacyclohexane ≥90.0% (by GC)
1,1,3,3,5,5-Hexaethoxy-1,3,5-trisilacyclohexane
Formel: C₁₅H₃₆O₆Si₃
Kogepunkt: 133°C
MDL Number: MFCD06660097
CAS-nummer: 17955-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ac788ec10a443358d1bd3b3450bb8879eac56d2&vid=3ac788ec10a443358d1bd3b3450bb8879eac56d2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR