1,1,2,3,3,3-Hexafluoropropyl methyl ether ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

H1507-5G H1507-25G
TCIAH1507-5GEA 550 DKK
TCIAH1507-5G TCIAH1507-25G
1,1,2,3,3,3-Hexafluoropropyl methyl ether ≥98.0% (by GC)
1,1,2,3,3,3-Hexafluoropropyl methyl ether
Formel: C₄H₄F₆O
Kogepunkt: 55°C
MDL Number: MFCD00041537
CAS-nummer: 382-34-3
UN nummer: 3271
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=419fbbf1955de35bb05aff7e8c26403f4b02a0da&vid=419fbbf1955de35bb05aff7e8c26403f4b02a0da&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR