2-Hydroxy-3-nitro-5-picolin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Methyl-3-nitro-2-pyridone, 2-Hydroxy-5-methyl-3-nitropyridine, 6-Hydroxy-5-nitro-3-picoline, 5-Methyl-3-nitro-2-pyridinol, 5-Methyl-3-nitro-2(1H)-pyridone

H1483-25G H1483-5G
TCIAH1483-25GEA 1800 DKK
TCIAH1483-25G TCIAH1483-5G
2-Hydroxy-3-nitro-5-picolin ≥98.0% (by GC, titration analysis)
2-Hydroxy-3-nitro-5-picolin
Formel: C₆H₆N₂O₃
MDL Number: MFCD02070021
CAS-nummer: 7464-14-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR