Hexaethylene glycol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H1432-5G
H1432-25G
TCIAH1432-5GEA
575
DKK
TCIAH1432-5G
TCIAH1432-25G
Hexaethylene glycol ≥98.0% (by GC)
Hexaethylene glycol
Formel:
C₁₂H₂₆O₇ Kogepunkt: 217°C Smeltepunkt: 7°C |
MDL Number:
MFCD00002877 CAS-nummer: 2615-15-8 EINECS: 220-045-1 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...