7-Hydroxycoumarin-3-carboxylic acid ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Umbelliferone-3-carboxylic Acid, 7-Hydroxy-2-oxo-2H-chromene-3-carboxylic acid

H1352-1G H1352-200MG
TCIAH1352-1GEA 1560 DKK
TCIAH1352-1G TCIAH1352-200MG
7-Hydroxycoumarin-3-carboxylic acid ≥98.0% (by HPLC)
7-Hydroxycoumarin-3-carboxylic acid
Formel: C₁₀H₆O₅
Smeltepunkt: 263°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00017491
CAS-nummer: 779-27-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR