(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(1-Hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile, 1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol

H1303-1G H1303-5G
TCIAH1303-1GEA 854 DKK
TCIAH1303-1G TCIAH1303-5G
(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile ≥98.0% (by HPLC, total nitrogen)
(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile
Formel: C₁₅H₁₉NO₂
Molekylvægt: 245.32 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06658142
CAS-nummer: 93413-76-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR