Hyaluronate tetrasaccharide

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

H1284-1MG H1284-5MG
TCIAH1284-1MGEA 1460 DKK
TCIAH1284-1MG TCIAH1284-5MG
Hyaluronate tetrasaccharide
Hyaluronate tetrasaccharide
Formel: C₂₈H₄₄N₂O₂₃
Opbevaringstemperatur: Fryser
CAS-nummer: 57282-61-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
TOF-MS to pass test
NMR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR