Hexyl methanesulphonate ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Methanesulphonic Acid Hexyl Ester
H1282-25G
H1282-5G
TCIAH1282-25GEA
1830
DKK
TCIAH1282-25G
TCIAH1282-5G
Hexyl methanesulphonate ≥96.0% (by GC)
Hexyl methanesulphonate
Formel:
C₇H₁₆O₃S Kogepunkt: 90°C |
MDL Number:
MFCD00674707 CAS-nummer: 16156-50-6 |
Specifikationer Testresultater
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...