D-Histidine methyl ester dihydrochloride ≥97.0% (by total nitrogen basis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
(R)-Methyl 2-amino-3-(1H-imidazol-4-yl)propanoate dihydrochloride, H-D-His-OMe.2HCl
H1213-1G
H1213-5G
TCIAH1213-1GEA
230
DKK
TCIAH1213-1G
TCIAH1213-5G
D-Histidine methyl ester dihydrochloride ≥97.0% (by total nitrogen basis)
D-Histidine methyl ester dihydrochloride
Formel:
C₇H₁₁N₃O₂ Molekylvægt: 242.10 g/mol Smeltepunkt: 197°C |
MDL Number:
MFCD00066137 CAS-nummer: 4467-54-3 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Specific rotation [a]20/D | -7.0 to -11.0 deg (C=2, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...