5-Hydroxy-1-indanone ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H1051-25G H1051-5G
TCIAH1051-25GEA 2470 DKK
TCIAH1051-25G TCIAH1051-5G
5-Hydroxy-1-indanone ≥98.0% (by GC, titration analysis)
5-Hydroxy-1-indanone
Formel: C9H8O2
Molekylvægt: 148.1600 g/mol
MDL Number: MFCD00857527
CAS-nummer: 3470-49-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 183.0 to 189.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR