Ethyl-2-(hydroxymethyl)acrylate ≥95.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-(Hydroxymethyl)acrylic acid ethyl ester, Ethyl 2-(hydroxymethyl)acrylate

Stabilization: stabiliseret med hydroquinon

H0916-25G H0916-5G
TCIAH0916-25GEA 2410 DKK
TCIAH0916-25G TCIAH0916-5G
Ethyl-2-(hydroxymethyl)acrylate ≥95.0% (by GC) stabilised
Ethyl-2-(hydroxymethyl)acrylate
Formel: C₆H₁₀O₃
Kogepunkt: 80°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01673859
CAS-nummer: 10029-04-6
UN nummer: 2810
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR